Please login to your online account to display your discounted pricing

CP 154526, Tocris

Selective, non-peptide CRF1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material CP 154526
Assay Percent Range >99%
Molecular Formula C23H32N4·HCl
Formula Weight 400.99
Synonym N-Butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

277910

 
r & d systems
277910
10mg Each for $252.39

277950

 
r & d systems
277950
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 400.99
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material CP 154526
Molecular Formula C23H32N4·HCl
Synonym N-Butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
CAS 257639-98-8
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C