Please login to your online account to display your discounted pricing

CP 135807, Tocris™

Selective 5-HT1D agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CP 135807
Assay Percent Range >98%
Molecular Formula C19H21N5O2
Formula Weight 351.4
Synonym 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-N-(3-nitro-2-pyridinyl)-1H-indol-5-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

378310

 
r & d systems
RDS378310
10mg Each for $239.63

378350

 
r & d systems
RDS378350
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 351.4
Quantity 10mg
Chemical Name or Material CP 135807
Synonym 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-N-(3-nitro-2-pyridinyl)-1H-indol-5-amine
Recommended Storage Store at +4°C
CAS 151272-90-1
Assay Percent Range >98%
Molecular Formula C19H21N5O2
Safety and Handling

Store at +4°C