Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ COX2, Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in Immunohistochemistry experiments with Thermo Scientific™ COX2, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

$301.37 - $1,191.94

Specifications

Antigen COX2
Clone SP21
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RM9121R7

 
thermo scientific™ lab vision™
RM9121R7
7mL Each for $690.88

RM9121S

 
thermo scientific™ lab vision™
RM9121S
1mL Each for $1,191.94

RM9121S0

 
thermo scientific™ lab vision™
RM9121S0
100µL Each for $301.37

RM9121S1

 
thermo scientific™ lab vision™
RM9121S1
500µL Each for $824.63
Description & Specifications

Specifications

Antigen COX2
Clone SP21
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Enzyme
Type Primary
Immunogen A synthetic peptide from C-terminus of rat cox2

Obtain accurate, reproducible results in Immunohistochemistry experiments with Thermo Scientific™ COX2, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.

Host Species: Rabbit

Clone: SP21

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of rat COX2

Molecular Weight: 70 kDa

Positive Control: Lung and colon CAs

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)