Please login to your online account to display your discounted pricing

COX IV Mouse anti-Human, eFluor 570, Clone: IB52C31H10, eBioscience™

Mouse Monoclonal Antibody

$275.88 - $448.05

Specifications

Antigen COX IV
Clone IB52C31H10
Host Species Mouse
Gene Alias Cytochrome c oxidase, Subunit IV, Complex IV
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123653

 
affymetrix
41977582
100μg Each for $448.05

501130709

 
affymetrix
41977580
25μg Each for $275.88
Description & Specifications

Specifications

Antigen COX IV
Clone IB52C31H10
Host Species Mouse
Gene Alias Cytochrome c oxidase, Subunit IV, Complex IV
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The IB52C31H10 monoclonal antibody recognizes human COX IV. COX IV, also known as cytochrome c oxidase (COX) or Complex IV, is the terminal complex of the electron transport chain. COX IV is encoded by nuclear DNA and localized to the inner mitochondrial membrane. COX IV functions in energy metabolism and shuttles electrons from cytochrome c to oxygen. Dysfunction in COX IV is associated with increased production of reactive oxygen species and cellular toxicity. Genetic defects in COX IV results in severe, often fatal metabolic disorders that primarily affect tissues with high energy demands such as heart, brain, and muscle.