missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

COX IV Mouse anti-Human, Alexa Fluor 488, Clone: IB52C31H10, eBioscience™

Mouse Monoclonal Antibody

$174.24 - $415.80

Specifications

Antigen COX IV
Clone IB52C31H10
Host Species Murine
Gene Alias Cytochrome c oxidase, Subunit IV, Complex IV
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124643 Life Technologies
LS53977580
25μg Each for $174.24
501124644 Life Technologies
LS53977582
100μg Each for $415.80
Specifications

Description

The IB52C31H10 monoclonal antibody recognizes human COX IV. COX IV, also known as cytochrome c oxidase (COX) or Complex IV, is the terminal complex of the electron transport chain. COX IV is encoded by nuclear DNA and localized to the inner mitochondrial membrane. COX IV functions in energy metabolism and shuttles electrons from cytochrome c to oxygen. Dysfunction in COX IV is associated with increased production of reactive oxygen species and cellular toxicity. Genetic defects in COX IV results in severe, often fatal metabolic disorders that primarily affect tissues with high energy demands such as heart, brain, and muscle.
Description & Specifications

Specifications

COX IV
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
IB52C31H10
Cytochrome c oxidase, Subunit IV, Complex IV
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Microscopy
Alexa Fluor 488
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL