Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ COX-1, Rabbit Polyclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ COX-1, Rabbit Polyclonal Antibody.

$237.16 - $1,198.20

Specifications

Antigen COX-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Format Quantity Price Quantity    

RB10687P

 
thermo scientific™ lab vision™
RB10687P
Concentrated 1mL Each for $1,198.20

RB10687P0

 
thermo scientific™ lab vision™
RB10687P0
Concentrated 100µL Each for $237.16

RB10687P1

 
thermo scientific™ lab vision™
RB10687P1
Concentrated 500µL Each for $752.10

RB10687R7

 
thermo scientific™ lab vision™
RB10687R7
Prediluted 7mL Each for $595.06
Description & Specifications

Specifications

Antigen COX-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Synthetic peptide corresponding to C-terminal of human COX-1
Purification Method Esophagus

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ COX-1, Rabbit Polyclonal Antibody.

Cyclooxygenases 1 and 2 (COX-1, COX-2) are enzymes involved in the conversion of arichidonate to prostaglandins. COX-1 and COX-2 are very similar in structure and function, though they vary in expression. COX-1 is normally expressed in most cell types, whereas COX-2 expression is at low levels unless induced by hormonal stimuli. In some breast cancer and bladder carcinoma studies, COX-1 was expressed in the stroma cells adjacent to the tumors but not in tumor cells.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminal of human COX-1

Molecular Weight: 72 kDa

Positive Control: Esophagus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO