missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Covidien Monoject™ SoftPack Hypodermic Needles

Color-coded gauge stripe and bar code on individual packages make it easy to identify needle sizes

$36.32 - $36.32

Specifications

For Use With (Application) Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Includes Polypropylene hub
Latex Free Yes
Material Stainless steel
Needle Point Style Regular Bevel
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Needle Length (Metric) Needle Needle Gauge Price Quantity  
Catalog Number Mfr. No. Length (English) Needle Length (Metric) Needle Needle Gauge Price Quantity  
22-557-172 Covidien
1188830340
0.75 in. 19.05mm 30 ga. Pack of 100 Each for $36.32
Case of 10 Pack for $363.20
Specifications

Description

  • Sterile, Precision-ground, ultra-sharp, tri-beveled, anti-coring surgical-grade stainless steel needles in a variety of sizes
  • Improved special-grade silicone needle lubricant makes injections smoother and less painful for patient
  • Translucent, color-coded luer lock hub helps identify needle gauge
  • Regular (A) Bevel needle points for intramuscular and subcutaneous injections

Description & Specifications

Specifications

Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Yes
Regular Bevel
Monoject™ Standard Hypodermic Needle
Polypropylene hub
Stainless steel
Sterile