Please login to your online account to display your discounted pricing

Covidien Monoject™ Rigid Pack 12mL Syringes

Convenient single-use individual packaging

$71.39 - $71.39

Specifications

Autoclavable Yes
Volume (Metric) 12mL
Disposable Yes
For Use With (Application) For use in O.R./procedure packs
Graduations 0.2mL
View More Specs
Products
Catalog Number Mfr. No. Termination Type Price Quantity    

22-652-090

 
covidien
8881512878
Luer-lock Monoject™ Syringe (no needle) Pack of 80 Each for $71.39
Case of 6 Pack for $428.34
Description & Specifications

Specifications

Autoclavable Yes
Volume (Metric) 12mL
Disposable Yes
For Use With (Application) For use in O.R./procedure packs
Graduations 0.2mL
Latex Free Yes
Sterile Yes

  • Autoclavable in rigid pack for use in O.R./procedure packs
  • Sterile    
  • Latex-free