Please login to your online account to display your discounted pricing

Covidien Monoject™ Hypodermic Needles with Aluminum Hub

Color-coded, two-piece rigid pack sheath coupled with a precision aluminum hub

$728.12 - $728.12

Specifications

Autoclavable Yes
For Use With (Application) Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Includes Precision aluminum hub
Latex Free Yes
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Gauge Length (English) Needle Length (Metric) Needle Needle Point Style Price Quantity    

22-257-161

 
covidien
8881200011
14 ga 1.5 in. 38.1mm Bevel Case of 1000 Each for $728.12
Description & Specifications

Specifications

Autoclavable Yes
For Use With (Application) Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Includes Precision aluminum hub
Latex Free Yes
Material Stainless Steel
Sterile Yes
Type Hypodermic Needle

  • Ultra-sharp tri-beveled surgical grade stainless steel coated with a special grade of silicone reduces “drag” on the injection site
  • Color-coded needle sheath allows for easy identification of needle gauge
  • Needles are autoclavable, allowing convenient use in hospital-prepared surgical packs
  • Cap-to-sheath seal provides visible and audible tamper evidence and guarantees sterility
  • Rigid polypropylene packaging protects against accidental product contamination