missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Covidien Autodrop™ Sharp Container for Phlebotomy
Fits into blood drawing trays
Manufacturer: Covidien 8901SA*
Description
- Portable
- Locks for final disposal
- Exterior dimensions: 4.25H × 4.25D × 4.25 in.W (10.8 × 10.8 × 10.8cm)
- Color: Red
Specifications
0.5 L | |
4.25 x 4.25 x 4.25 in. (10.8 x 10.8 x 10.8 cm) | |
10.8 cm | |
1 pt. | |
4.25 in. | |
4.25 in. |
4.25 in. | |
10.8 cm | |
10.8 cm | |
Red | |
4.25 in. |