missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coumarin, 99+%, ACROS Organics™

Manufacturer:  Acros Organics 110532500

 View more versions of this product

Catalog No. AC110532500



Specifications

Specifications

Coumarin
91-64-5
C9H6O2
17, 328
2H-1-Benzopyran-2-one
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
chromen-2-one
323
146.15
≥99.0%
68°C to 71°C
250g
99+%
99+%
MFCD00006850
15, 2550
Authentic
C1=CC=C2C(=C1)C=CC(=O)O2
146.145
CHEBI:28794
Crystals or Crystalline Powder
Plastic bottle
White

Chemical Identifiers

91-64-5
146.145
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
323
chromen-2-one
C9H6O2
MFCD00006850
2H-1-Benzopyran-2-one
CHEBI:28794
C1=CC=C2C(=C1)C=CC(=O)O2
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-086-7

RTECSNumber : GN4200000

TSCA : TSCA

SDS Documents