missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coumarin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A153360I

 View more versions of this product

Catalog No. AAA153360I



Description

Description

Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Coumarin
68°C to 71°C
297°C to 299°C
Pleasant
MFCD00006850
UN2811
14,2562
Soluble in water.
O=C1OC2=CC=CC=C2C=C1
146.15
CHEBI:28794
98%
91-64-5
0.935
162°C (323°F)
C9H6O2
5000 g
383644
coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
chromen-2-one
323
146.15

Chemical Identifiers

91-64-5
146.15
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
323
O=C1OC2=CC=CC=C2C=C1
C9H6O2
MFCD00006850
coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
CHEBI:28794
Safety and Handling

Safety and Handling

GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-086-7

RTECSNumber : GN4200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only