missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coumarin 6H, TCI America™

Manufacturer:  TCI America C2836200MG

 View more versions of this product

Catalog No. C2836200MG



Specifications

Specifications

Coumarin 6H
130°C
58336-35-9
MFCD00227485
MZSOXGPKUOAXNY-UHFFFAOYSA-N
241.29
241.29
≥97.0% (HPLC)
Yellow
200mg
C15H15NO2
2,3,6,7-Tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, 2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin
C1CC2=C3C(=C4C(=C2)C=CC(=O)O4)CCCN3C1
94022
Crystal-Powder at 20°C

Chemical Identifiers

58336-35-9
241.29
MZSOXGPKUOAXNY-UHFFFAOYSA-N
94022
C15H15NO2
MFCD00227485
2,3,6,7-Tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, 2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin
C1CC2=C3C(=C4C(=C2)C=CC(=O)O4)CCCN3C1
Safety and Handling

Safety and Handling

RTECSNumber : DJ3310000

TSCA : Yes

Documents