missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Coumarin-6-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H5476714

 View more versions of this product

Catalog No. AAH5476714



Description

Description

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Specifications

Coumarin-6-sulfonyl chloride
25g
C9H5ClO4S
UN3261
2-oxo-2h-1-benzopyran-6-sulfonyl chloride
HQIPMBGUDSOVEA-UHFFFAOYSA-N
2-oxochromene-6-sulfonyl chloride
2735833
97%
Moisture sensitive
10543-42-7
MFCD01941320
196167
Slightly soluble in water.
C1=CC2=C(C=CC(=O)O2)C=C1S(=O)(=O)Cl
244.645
244.65

Chemical Identifiers

10543-42-7
244.645
HQIPMBGUDSOVEA-UHFFFAOYSA-N
2735833
C1=CC2=C(C=CC(=O)O2)C=C1S(=O)(=O)Cl
C9H5ClO4S
MFCD01941320
2-oxo-2h-1-benzopyran-6-sulfonyl chloride
2-oxochromene-6-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents