missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coumarin-3-carboxylic acid, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC346765000

 View more versions of this product

Catalog No. AC346765000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Coumarin-3-carboxylic acid
Glass bottle
500g
98%
MFCD00006852
15, 2551
ACMLKANOGIVEPB-UHFFFAOYSA-N
2-oxochromene-3-carboxylic acid
10752
Crystalline Powder
Authentic
Brown
531-81-7
C10H6O4
18, 429
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O
190.154
190.15
≥97.5%

Chemical Identifiers

531-81-7
190.154
ACMLKANOGIVEPB-UHFFFAOYSA-N
10752
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O
C10H6O4
MFCD00006852
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van
2-oxochromene-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents