Please login to your online account to display your discounted pricing

Coumarin-3-carboxylic acid, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346761000

 View more versions of this product

Catalog No. AC346761000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 190.15
Quantity 100g
Color Light brown
Beilstein 18, 429
Merck Index 15, 2551
Chemical Name or Material Coumarin-3-carboxylic acid
Assay Percent Range 98%
Molecular Formula C10H6O4
CAS 531-81-7
Infrared Spectrum Authentic
MDL Number MFCD00006852
Packaging Glass bottle
Synonym 1-Oxo-2H-benzopyran-3-carboxylic acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Coumarin-3-carboxylic acid
CAS 531-81-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H6O4
Formula Weight 190.15
MDL Number MFCD00006852
Synonym 1-Oxo-2H-benzopyran-3-carboxylic acid
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents