missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Coumalic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2552114
Description
Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCoumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Coumalic acid | |
∼208°C (decomposition) | |
C6H4O4 | |
25 g | |
14,2557 | |
Very soluble in water. | |
OC(=O)C1=COC(=O)C=C1 | |
140.09 | |
140.1 |
500-05-0 | |
218°C (120 mmHg) | |
MFCD00006644 | |
119747 | |
coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g | |
ORGPJDKNYMVLFL-UHFFFAOYSA-N | |
2-oxo-2H-pyran-5-carboxylic acid | |
68141 | |
97% |
Chemical Identifiers
500-05-0 | |
140.09 | |
ORGPJDKNYMVLFL-UHFFFAOYSA-N | |
68141 |
C6H4O4 | |
MFCD00006644 | |
coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g | |
OC(=O)C1=COC(=O)C=C1 |
Safety and Handling
EINECSNumber : 207-899-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only