Please login to your online account to display your discounted pricing

Coulomat A, AQUASTAR™, EMD Millipore

$420.97 - $686.14

Chemical Identifiers

Chemical Name or Material Coulomat A
Name Note Xylene Blend
CAS 7553-56-2
Physical Form Liquid
Synonym Coulometric Anode Solution
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MAX1697AX4

 
emd millipore corporation
AX1697AX4
1L Each for $686.14

MAX1697AN4

 
emd millipore corporation
AX1697AN4
1L Each for $420.97
Description & Specifications

Specifications

Quantity 1L
Color Colorless to light yellow
Physical Form Liquid
CAS 7553-56-2
Name Note Xylene Blend
Chemical Name or Material Coulomat A
Recommended Storage Room Temperature
Synonym Coulometric Anode Solution
Safety and Handling

Room Temperature