Please login to your online account to display your discounted pricing

Coulomat A, AQUASTAR™, EMD Millipore


Quantity 1L

Manufacturer: emd millipore corporation  AX1697AN4

 View more versions of this product

Catalog No. MAX1697AN4

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Color Colorless to light yellow
Physical Form Liquid
Name Note Xylene Blend
Synonym Coulometric Anode Solution
Chemical Name or Material Coulomat A
Recommended Storage Room Temperature
CAS 7553-56-2

Specifications

Chemical Name or Material Coulomat A
Name Note Xylene Blend
CAS 7553-56-2
Physical Form Liquid
Synonym Coulometric Anode Solution
Accessories

Room Temperature