missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Superior Uniform Cotton Twill Knee-Length Lab Coats
100% cotton
Manufacturer: Superior Uniform 42342
Description
41 in. length based on size 40, 100% Sanforized Cotton Heavyweight Twill
420 Series
- Button front
- Left breast pocket
- Two lower outside pockets
- Side slits for pocket access
- Cloth knot buttons
- Left breast pocket
- Two lower outside pockets
- Side slits for pocket access
- Two-piece attached back belt with button closure
Order Info
Coats with cloth knot buttons and two-piece back belt also available. Call your Customer Service Representative or order online at www.fishersci.com.
Specifications
Sanforized Cotton Heavyweight Twill | |
Buttons | |
Cotton | |
42 | |
Men's |
2-piece back belt | |
White | |
3 | |
Cloth Knot Button |