missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Superior Uniform Cotton Twill Knee-Length Lab Coats

100% cotton

Supplier:  Superior Uniform 42340

 View more versions of this product

Catalog No. 01-369-3E



Description

Description

41 in. length based on size 40, 100% Sanforized Cotton Heavyweight Twill

420 Series

  • Button front
  • Left breast pocket
  • Two lower outside pockets
  • Side slits for pocket access
423 Series
  • Cloth knot buttons
  • Left breast pocket
  • Two lower outside pockets
  • Side slits for pocket access
  • Two-piece attached back belt with button closure

Order Info

Coats with cloth knot buttons and two-piece back belt also available. Call your Customer Service Representative or order online at www.fishersci.com.

Specifications

Specifications

40
Cotton
Buttons
3
41 in.
Sanforized Cotton Heavyweight Twill
Men's
White
2-piece back belt
Cloth Knot Button
104.14 cm
SDS
Documents

Documents

Promotions

Promotions