Please login to your online account to display your discounted pricing

Costunolide, Tocris

Inhibitor of human telomerase activity

$110.80 - $231.77

Chemical Identifiers

Chemical Name or Material Costunolide
Assay Percent Range >97%
Molecular Formula C15H20O2
Formula Weight 232.32
Synonym (3aS,6E,10E,11aR)-3a,4,5,8,9,11a-Hexahydro-6,10-dimethyl-3-methylene-cyclodeca[b]furan-2(3H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

248310

 
r & d systems
RDS248310
10mg Each for $231.77

24831

 
r & d systems
RDS24831
1mg Each for $110.80
Description & Specifications

Specifications

Formula Weight 232.32
Quantity 10mg
Chemical Name or Material Costunolide
Assay Percent Range >97%
CAS 553-21-9
Synonym (3aS,6E,10E,11aR)-3a,4,5,8,9,11a-Hexahydro-6,10-dimethyl-3-methylene-cyclodeca[b]furan-2(3H)-one
Recommended Storage Store at -20°C
Molecular Formula C15H20O2
Safety and Handling

Store at -20°C