missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ Transwell™ Inserts and Dishes

Applications include co-culture, drug transport, in vitro fertilization, and microbial pathogenesis studies.

Manufacturer:  Corning™ C3420

 View more versions of this product

Catalog No. 07-200-173



Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm thick translucent membrane
  • Pore sizes ranging from 0.4 µm to 8 µm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
Specifications

Specifications

Clear
44 cm2
Sterile
Cell Culture
13 mL (Well)
1/Pk.
9 mL
Round
75 mm
Polycarbonate
Insert
12/Cs.
3.0 μm
3 μm
SDS
Documents