missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ Stripettor™ Pipet Filler/Dispenser Replacement Parts

For use with Stripettor™ filler/dispenser

Manufacturer:  Corning™ C4095

 View more versions of this product

Catalog No. 07-201-792



Description

Description

  • PTFE filter (pack of 5) is for the Stripettor™ Plus pipetting controller
  • Pore Size: 0.45 μm
  • Autoclavable
Specifications

Specifications

Autoclavable
0.45μm
PTFE
Costar™ stripettor pipet filler/dispenser
Replacement Filter, 0.45μm
SDS
Documents
Product Certifications