Corning™ Costar™ Stripettor™ Pipet Filler/Dispenser Replacement Parts
For use with Stripettor™ filler/dispenser
Manufacturer:  Corning™ C4096
 View more versions of this product
Catalog No. 07-201-793
Description
Description
  • PTFE filter (pack of 5) is for the Stripettor™ Plus pipetting controller
  • Pore Size: 0.45 μm
  • Autoclavable
Specifications
Specifications
Not Autoclavable
Costar™ stripettor pipet filler/dispenser
Silicone
Pipet Holder
Documents