Please login to your online account to display your discounted pricing
Corning™ Costar™ Netwell™ Plates
$876.69 - $1,076.13
Specifications
Lid Yes
For Use With (Application) Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Color Clear
Sterile Yes
Well Color Clear
View More Specs
Products
Catalog Number Mfr. No. Format Type Price Quantity    
07-200-212
corning™
C3478
500mm mesh size, 12-well 12-Well NetWell™ Insert Case of 48 Each for $876.69
07-200-211
corning™
C3477
74μm mesh size, 12-well 12-Well NetWell™ Insert Case of 48 Each for $913.72
07-200-213
corning™
C3479
74mm mesh size, 6-well 6-Well NetWell™ Insert Case of 48 Each for $1,076.13
07-200-214
corning™
C3480
500mm mesh size, 6-well 6-Well NetWell™ Insert Case of 48 Each for $1,076.13
Description & Specifications
Specifications
Lid Yes
For Use With (Application) Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Color Clear
Sterile Yes
Well Color Clear
Well Design Round
Corning™ Netwell™ Plates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.