Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
Allows simultaneous processing
Manufacturer:  Corning™ C3520
 View more versions of this product
Catalog No. 07-200-223
Description
Description
  • These specially designed Netwell carriers and handles allow simultaneous processing of up to 12 Netwell inserts per carrier
  • Each carrier kit contains eight carriers and eight handles
  • Specifications
    Specifications
    Carrier Kit
    15 mm
    8/Cs.
    Allows simultaneous processing of up to (12) samples per carrier
    8/Pk.