Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
Manufacturer:  Corning™ C3208
 View more versions of this product
Catalog No. 07-200-185
Description
Description
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling.
  • Withstands a maximum RCF of 17,000 x g.
  • Costar low binding microcentrifuge tubes feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in better sample recovery.
  • RNase-/DNase-free.
Specifications
Specifications
Snap Cap
RNase, DNase-free
Polypropylene
Autoclavable
Natural
17000xG
1000/Pk.
Microcentrifuge Tube
Non-sterile
Natural
Snap Cap
0.65 mL
Polypropylene
500/Pk.
Conical
SDS
Documents
Accessories