missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ HTS Transwell™ -96 Well Permeable Support System

Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.

Manufacturer:  Corning™ C3391

 View more versions of this product

Catalog No. 07-200-741



Description

Description

  • The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm, and 8.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
  • 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
  • Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays.
  • Sterile
  • The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
  • The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
Specifications

Specifications

Standard
0.23 mL
Clear
4.26 mm
Polycarbonate
5/Cs.
5/Cs.
Sterile
8 x 12
With Lid
Flat
0.143 cm2
HTS Transwell™ unit (1 integral tray with 96 wells), reservoir plate (for cell growth steps), receiver plate (96 wells for growth or assay steps) and 2 sterile lids
TC-treated polycarbonate
0.4 μm
5/Pk.
96
SDS
Documents