Costar™ Flat Bottom Cell Culture Plates
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
$722.86 - $1,588.11
Products 4
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
07-200-87 View Documents Corning™
C3585
Case of 50 Each for $722.86
07-200-89 View Documents Corning™
C3595
Case of 50 Each for $793.60
07-200-91 View Documents Corning™
C3598
Case of 100 Each for $1,263.33
07-200-92 View Documents Corning™
C3599
Case of 100 Each for $1,588.11
Description
Description
  • Designed for a wide range of applications, including general cell growth experiments, cloning studies, virus isolation and susceptibility testing, and in vitro testing
  • Plate bottoms feature uniform thickness allowing for distortion-free observation and microscopic examination
  • Alphanumeric coordinates placed on the same focal plane as cell growth for convenient referencing of well position
  • Gripping edges make handling easier
  • All plates have nonreversible lids with condensation rings to minimize evaporation and risk of contamination
  • Made of optically clear, medical-grade polystyrene 
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic