missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles
Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions
Manufacturer: Corning™ C8388
Description
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Costar™ traditional style has smooth sides
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Nonpyrogenic
Specifications
HDPE | |
Plug Seal | |
4.2 oz., 125 mL | |
4.2 oz. | |
Polystyrene | |
1/Pk. | |
Red |
45 mm | |
Cylindrical | |
125 mL | |
Clear | |
24/Cs. | |
Sterile | |
45 |