missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ Disposable Aspirating Pipets

Constructed of polystyrene for strength, clarity, and durability. Corning™ Costar™ Disposable Aspirating Pipets are ideal for aseptically aspirating liquids by using vacuum suction.

Manufacturer:  Corning™ C9099

 View more versions of this product

Catalog No. 07-200-20



Description

Description

 

  • One piece polystyrene construction
  • Sterile and certified nonpyrogenic
  • No graduations; unplugged
  • Tip shape: round

 

Specifications

Specifications

Aspirating
13.7 in.
Round
34.8 cm
5 mL
Yes
Individually Wrapped, clear plastic wrap
0.169 oz.
Polystyrene
SDS
Documents
Product Certifications