missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Manufacturer: Corning™ C4412
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.
Specifications
Autoclavable | |
Polypropylene | |
Non-sterile | |
12/Pk. | |
Translucent | |
Rack | |
Strip | |
96 |
12/Rack | |
Round | |
8-Strip Microplate Cluster Tube | |
120/Cs. | |
1 mL | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
0.033 oz. |