missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 96-Well Cluster Tube System

Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.

Manufacturer:  Corning™ C4411

 View more versions of this product

Catalog No. 07-200-320



Description

Description

  • Composed of 96 polypropylene tubes in a standard microplate format.
  • Features 1.2 mL tubes that are available individually or in strips of eight tubes.
  • Polyethylene tube caps are available in 8-cap strips.
Specifications

Specifications

Autoclavable
Polypropylene
Sterile
96/Pk.
Translucent
Rack
Tube
96
96/Rack
Round
Microplate Cluster Tube
960/Cs.
1 mL
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges
0.033 oz.
SDS
Documents