Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Manufacturer:  Corning™ C3628
 View more versions of this product
Catalog No. 07-200-706
Description
Description
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
Flat
0.32 cm2
Polystyrene
100/Cs.
Sterile
96
With Lid
Standard
Clear
Attachment and growth of anchorage-dependent cells
100/Cs.
20/Pk.
Cell Culture
8 x 12
75 to 200 μL
Flat