Please login to your online account to display your discounted pricing

Corning™ Costar™ Cell Culture Triangular Flasks


Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.

Manufacturer: corning™  C3276

 View more versions of this product

Catalog No. 07-200-66

  • / Case of 100 Each
 

Description & Specifications

Specifications

Neck Style Straight
Sterile Yes
Material Polystyrene
Volume (Metric) Working 90mL
Closure Material HDPE
Shape Triangular
Closure Color Orange
Graduated Yes
For Use With (Application) For optimal cell attachment and growth
Culture Area 75cm2
Surface Coating Tissue culture treated

 

  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened