Please login to your online account to display your discounted pricing

Corning™ Costar™ Cell Culture Triangular Flasks


Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.

Manufacturer: corning™  C3056

 View more versions of this product

Catalog No. 07-200-63

  • / Case of 200 Each
 

Description & Specifications

Specifications

Closure Color Orange
Sterile Yes
Shape Triangular
Closure Material HDPE
Volume (Metric) Working 10mL
Surface Coating Tissue culture treated
Material Polystyrene
Graduated Yes
For Use With (Application) For optimal cell attachment and growth
Culture Area 25cm2
Neck Style Angled

 

  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened