missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: With Low Evaporation Lid

Manufacturer:  Corning™ C3595

 View more versions of this product

Catalog No. 07-200-89



Description

Description

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Specifications

Flat
0.32 cm2
Polystyrene
50/Cs.
1/Pk.
Sterile
96
With Lid
1/Pk
Flat
Clear
0.64 cm
50/Cs.
TC-Treated
0.36 mL
Cell Culture
8 x 12
0.1 to 0.2 mL
Standard
SDS
Documents
Product Certifications