missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Low Evaporation Lid
Manufacturer: Corning™ C3585
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
0.1 to 0.2 mL | |
Flat | |
0.32 cm2 | |
Polystyrene | |
50/Cs. | |
5/Pk. | |
Sterile | |
96 |
With Lid | |
Flat | |
Clear | |
0.64 cm | |
50/Cs. | |
TC-Treated | |
0.36 mL | |
Cell Culture | |
8 x 12 |