Corning™ Costar™ Flat Bottom Cell Culture Plates
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
$331.46 - $1,161.33
Specifications
Color Clear
Sterile Yes
Lid Yes
Material Polystyrene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Culture Area Diameter (Metric) Well Format Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Culture Area Diameter (Metric) Well Format Volume (Metric) Price Quantity  
07-200-740 Corning™
C3527
1.9cm2 1.6cm 24-well 3.5mL Case of 100 Each for $852.21
07-200-80 Corning™
C3506
9.5cm2 3.5cm 6-well 16.8mL Case of 100 Each for $653.33
07-200-81 Corning™
C3512
3.8cm2 2.26cm 12-well 6.9mL Case of 100 Each for $782.87
07-200-82 Corning™
C3513
4cm2 2.26cm 12-well 6.9mL Case of 50 Each for $384.54
07-200-83 Corning™
C3516
9.5cm2 3.5cm 6-well 16.8mL Case of 50 Each for $331.46
07-200-84 Corning™
C3524
1.9cm2 1.6cm 24-well 3.4mL Case of 100 Each for $836.62
07-200-87 Corning™
C3585
0.32cm2 0.64cm 96-well 0.36mL Case of 50 Each for $528.62
07-200-86 Corning™
C3548
0.95cm2 1.13cm 48-well 1.6mL Case of 100 Each for $990.13
07-200-89 Corning™
C3595
0.32cm2 0.64cm 96-well 0.36mL Case of 50 Each for $580.36
07-200-91 Corning™
C3598
0.32cm2 0.64cm 96-well 0.36mL Case of 100 Each for $923.79
07-200-90 Corning™
C3596
0.32cm2 0.64cm 96-well 0.36mL Case of 50 Each for $580.44
07-200-92 Corning™
C3599
0.32cm2 0.64cm 96-well 0.36mL Case of 100 Each for $1,161.33
09-761-146 Corning™
C3526
1.9cm2 1.6cm 24-well 3.5mL Case of 50 Each for $418.41
Specifications
Description
  • Designed for a wide range of applications, including general cell growth experiments, cloning studies, virus isolation and susceptibility testing, and in vitro testing
  • Plate bottoms feature uniform thickness—allowing for distortion-free observation and microscopic examination
  • Alphanumeric coordinates placed on the same focal plane as cell growth—for convenient referencing of well position
  • Gripping edges make handling easier
  • All plates have nonreversible lids with condensation rings to minimize evaporation and risk of contamination
  • Made of optically clear, medical-grade polystyrene 
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
Certifications
Certified nonpyrogenic
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Clear
Yes
Yes
Polystyrene