missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: With Lid

Manufacturer:  Corning™ C3799

 View more versions of this product

Catalog No. 07-200-95



Description

Description

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile
Specifications

Specifications

U
0.32 cm2
Attachment and growth of anchorage-dependent cells
50/Cs.
TC-Treated
0.33 mL
Clear
96
With Lid
75 to 200 μL
Cell Culture Microplate
Round
Clear
0.64 cm
Polystyrene
50/Cs.
1/Pk.
Sterile
Cell Culture
8 x 12
330 μL
1/Pk., 50/Cs.
Standard
SDS
Documents