missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 96-Well Half-Area Plates

Saves on valuable reagents by reducing the amount of reagent required per well, while retaining the ability to be read in standard microplate readers. Corning™ 96-Well Half Area Microplates are an economical solution for all of your assay diagnostic needs and are available in different colors and treatments.

$619.49 - $1,604.21

Specifications

Coding Alphanumeric
Format 96-well half-area
Material Polystyrene
Volume (Metric) Well 190μL
For Use With (Equipment) Plate readers
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Sterile Surface Coating Price Quantity  
Catalog Number Mfr. No. Closure Color Sterile Surface Coating Price Quantity  
07-200-309 Corning™
C3688
Yes White Yes Tissue Culture Case of 100 Each for $1,229.59
07-200-340 Corning™
C3686
No Black No NBS™ Case of 100 Each for $1,604.21
07-200-334 Corning™
C3697
Yes Clear Yes Tissue Culture Case of 100 Each for $995.64
07-200-326 Corning™
C3693
No White No Non-Treated Case of 100 Each for $1,101.74
07-200-339 Corning™
C3695
No Clear No Non-Treated Case of 100 Each for $839.95
07-200-735 Corning™
C3875
Yes Black Yes Tissue Culture Case of 100 Each for $1,499.21
07-200-329 Corning™
C3694
No Black No Non-Treated Case of 100 Each for $1,036.72
07-200-37 Corning™
C3690
No Clear No High binding Case of 100 Each for $930.67
07-200-93 Corning™
C3696
Yes Clear Yes Tissue Culture Case of 50 Each for $619.49
Specifications

Description

 

  • Polystyrene
  • Flat wells
  • Surface area per well: 0.16cm2
  • Alphanumeric grid
  • Standard external dimensions compatible with most plate readers
  • Clear microplates are used for cell culture and colorimetric (absorbance) assays
  • Black and white microplates can be used for fluorescent and luminescent assays
    • Black reduces well-to-well crosstalk and background for fluorescent assays
    • White reduces well-to-well crosstalk to enhance luminescent signals and reduce background for luminescent assays
  • Choice of four surfaces: untreated for homogenous assays, high-binding for EIA applications, non-binding surface (NBS™) for the minimization of protein and nucleic acid binding, or tissue culture treated for cell growth

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Alphanumeric
Polystyrene
Plate readers
96
96-well half-area
190μL
25 to 125μL