Corning™ 96-Well Clear PVC Assay Microplates
Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.
Manufacturer:  Corning™ C2595
 View more versions of this product
Catalog No. 07-200-33
Description
Description
  • Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 μL (260 μL for V-bottom); working well volume of 50 μL to 150 μL
  • Lids are not available.
  • Specifications
    Specifications
    Standard
    50 to 150μL
    Clear
    PVC
    100/Cs.
    Non-sterile
    Non-Treated
    8 x 12
    Without Lid
    Flat
    Solution-based assays, serial dilutions and general storage applications
    100/Cs.
    25/Pk.
    Flat
    96