Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Manufacturer: Corning™ C9018
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
75 to 200 μL | |
Flat | |
Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions. | |
100/Cs. | |
High Bind | |
Non-sterile | |
High Binding | |
8 x 12 |
Without Lid | |
Flat | |
Clear | |
Polystyrene | |
100/Cs. | |
25/Pk. | |
High Bind | |
96 |