Corning™ 96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Manufacturer:  Corning™ C3922
 View more versions of this product
Catalog No. 07-200-591
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications
    Specifications
    Standard
    4 inner packs of 25
    360 μL
    Flat
    White
    Polystyrene
    100/Cs.
    Non-sterile
    96
    Without Lid
    EIA/RIA
    75 to 200 μL
    Flat
    Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
    100/Cs.
    25/Pk.
    High Bind
    8 x 12