Corning™ 96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Manufacturer: Corning™ C3912
Description
Specifications
Standard | |
75 to 200 μL | |
Flat | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
100/Cs. | |
25/Pk. | |
Non-treated | |
8 x 12 |
Without Lid | |
Flat | |
White | |
Polystyrene | |
100/Cs. | |
Non-sterile | |
96 |