missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.
Manufacturer: Corning™ C3631
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
4 inner packs of 25 | |
360 μL | |
Flat | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
100 | |
Non-sterile | |
96 |
Without Lid | |
EIA/RIA | |
75 to 200 μL | |
Black, Clear Bottom | |
Polystyrene | |
100/Cs. | |
Non-treated | |
8 x 12 |