missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid

Manufacturer:  Corning™ C3904

 View more versions of this product

Catalog No. 07-200-588



Description

Description

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications

    Specifications

    Flat
    0.32 cm2
    Polystyrene
    100/Cs.
    Sterile
    96
    With Lid
    20/Pk., 100/Cs.
    Standard
    Black, Clear Bottom
    Attachment and growth of anchorage-dependent cells
    100/Cs.
    20/Pk.
    Cell Culture
    8 x 12
    75 to 200 μL
    Assay Plate
    Flat
    SDS
    Documents