Corning™ 96-Well Assay Blocks
Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.
Manufacturer:  Corning™ C3961
 View more versions of this product
Catalog No. 07-200-701
Description
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile
Specifications
Specifications
8 x 12
V
For compound storage, assay and cell-growth application
100/Cs.
5/Pk.
Non-treated
Without Lid
20 inner packs of 5
Standard
Not Treated
Clear
Polypropylene
100/Cs.
Non-sterile
96
2000 μL
96-well Assay Blocks
V-bottom