Please login to your online account to display your discounted pricing

Alfa Aesar™ Cortistatin 29


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66998EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66998EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Product Type Cortistatin 29
Quantity 0.5mg
Format Solid
Regulatory Status RUO
Molecular Weight 3542.10
Purification Method HPLC